3-(3-Chlorophenyl)-3-fluoropyrrolidine
Lieferant: Apollo Scientific
PC510218-1G
APOSPC510218-1GEA
1220
EUR
APOSPC510218-1G
3-(3-Chlorophenyl)-3-fluoropyrrolidine
3-(3-Chlorophenyl)-3-fluoropyrrolidine
Formel:
C₁₀H₁₁ClFN Molecular Weight: 199,652 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26038927 CAS-Nummer: 1565489-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...