N-[[4-(4-Fluorobenzyl)-2-morpholinyl]methyl]acetamide

Lieferant: Apollo Scientific

Synonyme: N-((4-(4-Fluorobenzyl)morpholin-2-yl)methyl)acetamide

PC510236-100G PC510236-25G
APOSPC510236-100GEA 116 EUR
APOSPC510236-100G APOSPC510236-25G
N-[[4-(4-Fluorobenzyl)-2-morpholinyl]methyl]acetamide
N-[[4-(4-Fluorobenzyl)-2-morpholinyl]methyl]acetamide
Formel: C₁₄H₁₉FN₂O₂
Molecular Weight: 266.311 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619692
CAS-Nummer: 112913-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR