2-(Aminomethyl)-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC510237-1G
APOSPC510237-1GEA 751 EUR
APOSPC510237-1G
2-(Aminomethyl)-6-(trifluoromethyl)pyridine
2-(Aminomethyl)-6-(trifluoromethyl)pyridine
Formel: C₇H₇F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10696232
CAS-Nummer: 916304-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR