4-(Aminomethyl)-3-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific

PC510238-1G
APOSPC510238-1GEA 804 EUR
APOSPC510238-1G
4-(Aminomethyl)-3-(trifluoromethyl)pyrazole
4-(Aminomethyl)-3-(trifluoromethyl)pyrazole
Formel: C₅H₆F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28505588
CAS-Nummer: 1525641-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR