3-Methyl-2-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC510241-5G
APOSPC510241-5GEA 561 EUR
APOSPC510241-5G
3-Methyl-2-(trifluoromethyl)pyridine
3-Methyl-2-(trifluoromethyl)pyridine
Formel: C₇H₆F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD07774129
CAS-Nummer: 1620-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR