1-(2-Chloroethyl)-2-fluorobenzene

Lieferant: Apollo Scientific

PC510242-1G PC510242-250MG PC510242-5G
APOSPC510242-1GEA 478 EUR
APOSPC510242-1G APOSPC510242-250MG APOSPC510242-5G
1-(2-Chloroethyl)-2-fluorobenzene
1-(2-Chloroethyl)-2-fluorobenzene
Formel: C₈H₈ClF
Lagertemperatur: Raumtemperatur
MDL: MFCD11110026
CAS-Nummer: 119779-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR