1-(4-Amino-3-fluorophenyl)pyridin-2-one

Lieferant: Apollo Scientific

PC510244-1G
APOSPC510244-1GEA 601 EUR
APOSPC510244-1G
1-(4-Amino-3-fluorophenyl)pyridin-2-one
1-(4-Amino-3-fluorophenyl)pyridin-2-one
Formel: C₁₁H₉FN₂O
Molecular Weight: 204,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12974818
CAS-Nummer: 536747-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR