Methyl 1-Methyl-5-[4-(trifluoromethoxy)phenyl]pyrrole-2-carboxylate

Lieferant: Apollo Scientific

PC510246-1G
APOSPC510246-1GEA 651 EUR
APOSPC510246-1G
Methyl 1-Methyl-5-[4-(trifluoromethoxy)phenyl]pyrrole-2-carboxylate
Methyl 1-Methyl-5-[4-(trifluoromethoxy)phenyl]pyrrole-2-carboxylate
Formel: C₁₄H₁₂F₃NO₃
Molecular Weight: 299,245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28954614
CAS-Nummer: 2006276-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR