Methyl 1-Methyl-5-[4-(trifluoromethyl)phenyl]pyrrole-2-carboxylate
Lieferant: Apollo Scientific
PC510247-1G
APOSPC510247-1GEA
855
EUR
APOSPC510247-1G
Methyl 1-Methyl-5-[4-(trifluoromethyl)phenyl]pyrrole-2-carboxylate
Methyl 1-Methyl-5-[4-(trifluoromethyl)phenyl]pyrrole-2-carboxylate
Formel:
C₁₄H₁₂F₃NO₂ Molecular Weight: 283,246 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28954611 CAS-Nummer: 1895780-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...