3-Ethoxy-2-fluoropyridine

Lieferant: Apollo Scientific

PC510249-1G PC510249-250MG PC510249-5G
APOSPC510249-1GEA 794 EUR
APOSPC510249-1G APOSPC510249-250MG APOSPC510249-5G
3-Ethoxy-2-fluoropyridine
3-Ethoxy-2-fluoropyridine
Formel: C₇H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD18384511
CAS-Nummer: 847225-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR