5-Fluoro-2-(hydroxymethyl)-4-methylbenzimidazole

Lieferant: Apollo Scientific

PC510250-1G
APOSPC510250-1GEA 1010 EUR
APOSPC510250-1G
5-Fluoro-2-(hydroxymethyl)-4-methylbenzimidazole
5-Fluoro-2-(hydroxymethyl)-4-methylbenzimidazole
Formel: C₉H₉FN₂O
Molecular Weight: 180,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977366
CAS-Nummer: 1936068-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR