4-Bromo-6-fluoro-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

PC510251-1G
APOSPC510251-1GEA 916 EUR
APOSPC510251-1G
4-Bromo-6-fluoro-2-(hydroxymethyl)benzimidazole
4-Bromo-6-fluoro-2-(hydroxymethyl)benzimidazole
Formel: C₈H₆BrFN₂O
Molecular Weight: 245,048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977364
CAS-Nummer: 1935913-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR