4,6-Difluoro-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

PC510252-1G
APOSPC510252-1GEA 1010 EUR
APOSPC510252-1G
4,6-Difluoro-2-(hydroxymethyl)benzimidazole
4,6-Difluoro-2-(hydroxymethyl)benzimidazole
Formel: C₈H₆F₂N₂O
Molecular Weight: 184,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977363
CAS-Nummer: 1505508-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR