2-(Hydroxymethyl)-5-(trifluoromethoxy)benzimidazole

Lieferant: Apollo Scientific

PC510253-1G
APOSPC510253-1GEA 1010 EUR
APOSPC510253-1G
2-(Hydroxymethyl)-5-(trifluoromethoxy)benzimidazole
2-(Hydroxymethyl)-5-(trifluoromethoxy)benzimidazole
Formel: C₉H₇F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28977362
CAS-Nummer: 919054-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR