4,5-Difluoro-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

PC510254-1G
APOSPC510254-1GEA 672 EUR
APOSPC510254-1G
4,5-Difluoro-2-(hydroxymethyl)benzimidazole
4,5-Difluoro-2-(hydroxymethyl)benzimidazole
Formel: C₈H₆F₂N₂O
Molecular Weight: 184.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26034283
CAS-Nummer: 1517146-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR