5,6-Difluoro-2-(hydroxymethyl)-benzimidazole
Lieferant: Apollo Scientific
PC510255-1G
APOSPC510255-1GEA
916
EUR
APOSPC510255-1G
5,6-Difluoro-2-(hydroxymethyl)-benzimidazole
5,6-Difluoro-2-(hydroxymethyl)-benzimidazole
Formel:
C₈H₆F₂N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01628314 CAS-Nummer: 1344314-81-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...