2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole

Lieferant: Apollo Scientific

PC510256-5G
APOSPC510256-5GEA 0 EUR
APOSPC510256-5G
2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole
2-(Hydroxymethyl)-5-(trifluoromethyl)benzimidazole
Formel: C₉H₇F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD12755183
CAS-Nummer: 6758-34-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR