2-(3-Chloropropyl)-6-fluoroquinazoline

Lieferant: Apollo Scientific

PC510258-1G
APOSPC510258-1GEA 1010 EUR
APOSPC510258-1G
2-(3-Chloropropyl)-6-fluoroquinazoline
2-(3-Chloropropyl)-6-fluoroquinazoline
Formel: C₁₁H₁₀ClFN₂
Molecular Weight: 224,662 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977355
CAS-Nummer: 2006277-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR