2-(2-Chloroethyl)-6-fluoroquinazoline

Lieferant: Apollo Scientific

PC510259-1G
APOSPC510259-1GEA 1010 EUR
APOSPC510259-1G
2-(2-Chloroethyl)-6-fluoroquinazoline
2-(2-Chloroethyl)-6-fluoroquinazoline
Formel: C₁₀H₈ClFN₂
Molecular Weight: 210,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977354
CAS-Nummer: 1936487-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR