2-(Chloromethyl)-6-fluoroquinazoline

Lieferant: Apollo Scientific

PC510260-1G
APOSPC510260-1GEA 1010 EUR
APOSPC510260-1G
2-(Chloromethyl)-6-fluoroquinazoline
2-(Chloromethyl)-6-fluoroquinazoline
Formel: C₉H₆ClFN₂
Molecular Weight: 196,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977353
CAS-Nummer: 1935241-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR