Ethyl 4-Chloro-6-(trifluoromethyl)quinoline-3-carboxylate

Lieferant: Apollo Scientific

PC510261-1G
APOSPC510261-1GEA 753 EUR
APOSPC510261-1G
Ethyl 4-Chloro-6-(trifluoromethyl)quinoline-3-carboxylate
Ethyl 4-Chloro-6-(trifluoromethyl)quinoline-3-carboxylate
Formel: C₁₃H₉ClF₃NO₂
Molecular Weight: 303.664 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173370
CAS-Nummer: 193827-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR