2-Chloro-7-(trifluoromethyl)quinoline-3-carbaldehyde

Lieferant: Apollo Scientific

PC510263-1G
APOSPC510263-1GEA 916 EUR
APOSPC510263-1G
2-Chloro-7-(trifluoromethyl)quinoline-3-carbaldehyde
2-Chloro-7-(trifluoromethyl)quinoline-3-carbaldehyde
Formel: C₁₁H₅ClF₃NO
Molecular Weight: 259,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977409
CAS-Nummer: 1621615-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR