Ethyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

PC510265-1G
APOSPC510265-1GEA 1220 EUR
APOSPC510265-1G
Ethyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Ethyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Formel: C₁₄H₁₂F₃NO₄
Molecular Weight: 315,245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977400
CAS-Nummer: 2006278-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR