Methyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

PC510266-1G
APOSPC510266-1GEA 1220 EUR
APOSPC510266-1G
Methyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Methyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Formel: C₁₃H₁₀F₃NO₄
Molecular Weight: 301,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977399
CAS-Nummer: 2006277-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR