3-[[(2-Tetrahydropyranyl)oxy][4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
Lieferant: Apollo Scientific
PC510267-1G
APOSPC510267-1GEA
1220
EUR
APOSPC510267-1G
3-[[(2-Tetrahydropyranyl)oxy][4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
3-[[(2-Tetrahydropyranyl)oxy][4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
Formel:
C₁₇H₁₆F₃NO₅ Molecular Weight: 371,308 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977398 CAS-Nummer: 2006277-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...