3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
Lieferant: Apollo Scientific
PC510268-1G
APOSPC510268-1GEA
1220
EUR
APOSPC510268-1G
3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylic acid
Formel:
C₁₂H₈F₃NO₄ Molecular Weight: 287,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977397 CAS-Nummer: 2006277-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...