Ethyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

PC510269-1G
APOSPC510269-1GEA 1220 EUR
APOSPC510269-1G
Ethyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Ethyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formel: C₁₃H₁₂FNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD28977392
CAS-Nummer: 2006277-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR