Methyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

PC510270-1G
APOSPC510270-1GEA 1220 EUR
APOSPC510270-1G
Methyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Methyl 3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formel: C₁₂H₁₀FNO₄
Molecular Weight: 251,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977391
CAS-Nummer: 2006277-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR