3-[(4-Fluorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

PC510271-1G
APOSPC510271-1GEA 1220 EUR
APOSPC510271-1G
3-[(4-Fluorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Fluorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Formel: C₁₆H₁₆FNO₅
Molecular Weight: 321,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977390
CAS-Nummer: 2006277-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR