3-(3-Fluorophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

PC510274-5G
APOSPC510274-5GEA 1010 EUR
APOSPC510274-5G
3-(3-Fluorophenyl)-2-hydroxypropionic acid
3-(3-Fluorophenyl)-2-hydroxypropionic acid
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21296531
CAS-Nummer: 246137-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR