(S)-3-(4-Fluorophenyl)-2-hydroxypropionic acid

Lieferant: Apollo Scientific

PC510275-5G
APOSPC510275-5GEA 1010 EUR
APOSPC510275-5G
(S)-3-(4-Fluorophenyl)-2-hydroxypropionic acid
(S)-3-(4-Fluorophenyl)-2-hydroxypropionic acid
Formel: C₉H₉FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28977375
CAS-Nummer: 124980-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR