3-(2-Chloro-4-fluorophenyl)propanoic acid

Lieferant: Apollo Scientific

PC510277-5G
APOSPC510277-5GEA 734 EUR
APOSPC510277-5G
3-(2-Chloro-4-fluorophenyl)propanoic acid
3-(2-Chloro-4-fluorophenyl)propanoic acid
Formel: C₉H₈ClFO₂
Molecular Weight: 202,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656905
CAS-Nummer: 174603-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR