3-(Difluoromethoxy)-2-nitropyridine

Lieferant: Apollo Scientific
Achtung

PC510284-1G
APOSPC510284-1GEA 601 EUR
APOSPC510284-1G
3-(Difluoromethoxy)-2-nitropyridine
3-(Difluoromethoxy)-2-nitropyridine
Formel: C₆H₄F₂N₂O₃
Molecular Weight: 190,104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405787
CAS-Nummer: 1111637-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR