1,3-Dibromo-2-(difluoromethoxy)benzene

Lieferant: Apollo Scientific

Synonyme: 1,3-Dibromo-2-difluoromethoxy-benzene

PC510285-1G
APOSPC510285-1GEA 601 EUR
APOSPC510285-1G
1,3-Dibromo-2-(difluoromethoxy)benzene
1,3-Dibromo-2-(difluoromethoxy)benzene
Formel: C₇H₄Br₂F₂O
Molecular Weight: 301.911 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22418262
CAS-Nummer: 1182728-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR