2-Fluoro-6-methoxy-3-(methylsulphonyl)benzoic acid

Lieferant: Apollo Scientific

PC510291-5G
APOSPC510291-5GEA 1540 EUR
APOSPC510291-5G
2-Fluoro-6-methoxy-3-(methylsulphonyl)benzoic acid
2-Fluoro-6-methoxy-3-(methylsulphonyl)benzoic acid
Formel: C₉H₉FO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD29049241
CAS-Nummer: 2006277-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR