Methyl (R)-3-(Dibenzylamino)-2-fluoropropanoate

Lieferant: Apollo Scientific

PC510296-1G
APOSPC510296-1GEA 611 EUR
APOSPC510296-1G
Methyl (R)-3-(Dibenzylamino)-2-fluoropropanoate
Methyl (R)-3-(Dibenzylamino)-2-fluoropropanoate
Formel: C₁₈H₂₀FNO₂
Molecular Weight: 301,355 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29055298
CAS-Nummer: 88099-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR