(R)-4-(Dibenzylamino)-3-fluoro-2-methyl-2-butanol
Lieferant: Apollo Scientific
PC510297-1G
APOSPC510297-1GEA
611
EUR
APOSPC510297-1G
(R)-4-(Dibenzylamino)-3-fluoro-2-methyl-2-butanol
(R)-4-(Dibenzylamino)-3-fluoro-2-methyl-2-butanol
Formel:
C₁₉H₂₄FNO Molecular Weight: 301,398 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29055296 CAS-Nummer: 1609545-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...