3-Fluor-4-(4-methoxybenzyloxycarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

PC5103-1G
APOSPC5103-1GEA 139 EUR
APOSPC5103-1G
3-Fluor-4-(4-methoxybenzyloxycarbamoyl)phenylboronsäure 98%
3-Fluor-4-(4-methoxybenzyloxycarbamoyl)phenylboronsäure
Formel: C₁₅H₁₅BFNO₅
Schmelzpunkt: 150…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235097
CAS-Nummer: 913835-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR