3-Bromo-4-(difluoromethoxy)phenylacetic acid

Lieferant: Apollo Scientific

PC510304-1G
APOSPC510304-1GEA 1010 EUR
APOSPC510304-1G
3-Bromo-4-(difluoromethoxy)phenylacetic acid
3-Bromo-4-(difluoromethoxy)phenylacetic acid
Formel: C₉H₇BrF₂O₃
Molecular Weight: 281.051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29065908
CAS-Nummer: 2006277-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR