α,α-Difluoro-1-naphthaleneacetic acid

Lieferant: Apollo Scientific

PC510306-1G
APOSPC510306-1GEA 855 EUR
APOSPC510306-1G
α,α-Difluoro-1-naphthaleneacetic acid
α,α-Difluoro-1-naphthaleneacetic acid
Formel: C₁₂H₈F₂O₂
Molecular Weight: 222,188 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16756770
CAS-Nummer: 73790-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR