Ethyl 2,2-Difluoro-2-(1-naphthyl)acetate

Lieferant: Apollo Scientific

PC510307-1G
APOSPC510307-1GEA 804 EUR
APOSPC510307-1G
Ethyl 2,2-Difluoro-2-(1-naphthyl)acetate
Ethyl 2,2-Difluoro-2-(1-naphthyl)acetate
Formel: C₁₄H₁₂F₂O₂
Molecular Weight: 250,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17064708
CAS-Nummer: 73790-00-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR