Ethyl 2-(4-Acetylphenyl)-2,2-difluoroacetate

Lieferant: Apollo Scientific

PC510309-1G
APOSPC510309-1GEA 804 EUR
APOSPC510309-1G
Ethyl 2-(4-Acetylphenyl)-2,2-difluoroacetate
Ethyl 2-(4-Acetylphenyl)-2,2-difluoroacetate
Formel: C₁₂H₁₂F₂O₃
Molecular Weight: 242,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25957738
CAS-Nummer: 1308915-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR