2,2-Difluoro-2-mesitylacetic acid
Lieferant: Apollo Scientific
PC510312-1G
APOSPC510312-1GEA
855
EUR
APOSPC510312-1G
2,2-Difluoro-2-mesitylacetic acid
2,2-Difluoro-2-mesitylacetic acid
Formel:
C₁₁H₁₂F₂O₂ Molecular Weight: 214,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29065901 CAS-Nummer: 1436389-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...