Ethyl 2,2-Difluoro-2-mesitylacetate

Lieferant: Apollo Scientific

PC510313-1G
APOSPC510313-1GEA 804 EUR
APOSPC510313-1G
Ethyl 2,2-Difluoro-2-mesitylacetate
Ethyl 2,2-Difluoro-2-mesitylacetate
Formel: C₁₃H₁₆F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29065900
CAS-Nummer: 1436389-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR