2,2-Difluoro-2-(2-nitrophenyl)acetic acid

Lieferant: Apollo Scientific

PC510314-1G
APOSPC510314-1GEA 855 EUR
APOSPC510314-1G
2,2-Difluoro-2-(2-nitrophenyl)acetic acid
2,2-Difluoro-2-(2-nitrophenyl)acetic acid
Formel: C₈H₅F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD29065899
CAS-Nummer: 1308915-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR