Ethyl 2,2-Difluoro-2-(2-nitrophenyl)acetate

Lieferant: Apollo Scientific
Achtung

PC510315-1G
APOSPC510315-1GEA 804 EUR
APOSPC510315-1G
Ethyl 2,2-Difluoro-2-(2-nitrophenyl)acetate
Ethyl 2,2-Difluoro-2-(2-nitrophenyl)acetate
Formel: C₁₀H₉F₂NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD29065898
CAS-Nummer: 1308915-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR