Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate
Lieferant: Apollo Scientific
PC510317-1G
APOSPC510317-1GEA
1010
EUR
APOSPC510317-1G
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate
Ethyl 2-(4-Cyanophenyl)-2,2-difluoroacetate
Formel:
C₁₁H₉F₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD25967876 CAS-Nummer: 854778-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...