2-[4-(Carboxymethyl)phenyl]-2,2-difluoroacetic acid
Lieferant: Apollo Scientific
PC510318-1G
APOSPC510318-1GEA
1010
EUR
APOSPC510318-1G
2-[4-(Carboxymethyl)phenyl]-2,2-difluoroacetic acid
2-[4-(Carboxymethyl)phenyl]-2,2-difluoroacetic acid
Formel:
C₁₀H₈F₂O₄ Molecular Weight: 230,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29065897 CAS-Nummer: 1436389-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...