Ethyl 2-[4-(2-Ethoxy-2-oxoethyl)phenyl]-2,2-difluoroacetate
Lieferant: Apollo Scientific
PC510319-1G
APOSPC510319-1GEA
804
EUR
APOSPC510319-1G
Ethyl 2-[4-(2-Ethoxy-2-oxoethyl)phenyl]-2,2-difluoroacetate
Ethyl 2-[4-(2-Ethoxy-2-oxoethyl)phenyl]-2,2-difluoroacetate
Formel:
C₁₄H₁₆F₂O₄ Molecular Weight: 286,271 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29065896 CAS-Nummer: 2006277-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...