Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate

Lieferant: Apollo Scientific

PC510321-1G
APOSPC510321-1GEA 1010 EUR
APOSPC510321-1G
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate
Formel: C₁₁H₉F₂NO₂
Molecular Weight: 225,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16756772
CAS-Nummer: 1249788-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR